4-(Trifluoromethyl)thiophenol ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol

A12725.03 A12725.06 A12725.14
A12725.03EA 70.6 EUR
A12725.03 A12725.06 A12725.14
4-(Trifluoromethyl)thiophenol ≥97%
4-(Trifluoromethyl)thiophenol

Purity: 97% CAS No.: 825-83-2 Molecular Formula: C7H5F3S Formula Weight: 178.18 Refractive Index: 1.4920 Odour: Stench Boiling Point: 66-68°/15mm Beilstein Reference: 2045665 UN No.: 2810 MDL No.: MFCD00042320

Formula: C₇H₅F₃S
MW: 178.18 g/mol
Boiling Pt: 66…68 °C (15 mmHg)
Density: 1.300
Storage Temperature: Ambient
MDL Number: MFCD00042320
CAS Number: 825-83-2
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR