1-Dodecanethiol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A12741.22 A12741.36
A12741.22EA 26.9 EUR
A12741.22 A12741.36
1-Dodecanethiol ≥98%
1-Dodecanethiol

Purity: 98% CAS No.: 112-55-0 Molecular Formula: C12H26S Formula Weight: 202.40 Flash Point: 133°(271°F) Refractive Index: 1.4601 Odour: Unpleasant Melting Point: -7° Boiling Point: 276-278° Beilstein Reference: 969337 MDL No.: MFCD00004885

Formula: C₁₂H₂₆S
Boiling Pt: 276…278 °C
Melting Pt: –7 °C
Density: 0,845
Flash Pt: 133 °C (271 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004885
CAS Number: 112-55-0
EINECS: 203-984-1
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR