Methoxyhydroquinone 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Methoxyhydroquinone

A12847.09 A12847.18 A12847.30
A12847.09EA 44.2 EUR
A12847.09 A12847.18 A12847.30
Methoxyhydroquinone 97
Methoxyhydroquinone

Purity: 97% CAS No.: 824-46-4 Molecular Formula: C7H8O3 Formula Weight: 140.14 Melting Point: 87-91° Beilstein Reference: 2045285

Formula: C₇H₈O₃
MW: 140.14 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00013971
CAS Number: 824-46-4
EINECS: 212-530-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR