4-Bromobutyric acid ≥97%

Supplier: Thermo Fisher Scientific
Danger

A12870.09 A12870.18 A12870.30
A12870.09EA 55.2 EUR
A12870.09 A12870.18 A12870.30
4-Bromobutyric acid ≥97%
4-Bromobutyric acid

Purity: 97% CAS No.: 2623-87-2 Molecular Formula: Br(CH2)3CO2H Formula Weight: 167.01 Flash Point: 115°(239°F) Odour: Stench Boiling Point: 129-131°/11mm Beilstein Reference: 1743056 UN No.: 3261 MDL No.: MFCD00002817

Formula: BrH₂C(CH₂)₂COOH
Boiling Pt: 129…131 °C (11 mmHg)
Melting Pt: 31…33 °C
Flash Pt: 115 °C (239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002817
CAS Number: 2623-87-2
EINECS: 220-083-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR