Maleimide ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1H-Pyrrole-2,5-dione

A13135.18 A13135.06 A13135.14 A13135.30
A13135.18EA 363 EUR
A13135.18 A13135.06 A13135.14 A13135.30
Maleimide ≥98%
Maleimide

Purity: 98+% CAS No.: 541-59-3 Molecular Formula: C4H3NO2 Formula Weight: 97.07 Melting Point: 90-94° Boiling Point: 97-103/5mm Solubility: Soluble in water Beilstein Reference: 106910 UN No.: 2923 MDL No.: MFCD00005494

Formula: C₄H₃NO₂
MW: 97.07 g/mol
Boiling Pt: 97…103 °C (5 mmHg)
Melting Pt: 90…94 °C
Density: 1.249
Storage Temperature: Ambient
MDL Number: MFCD00005494
CAS Number: 541-59-3
EINECS: 208-787-4
UN: 2923
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR