trans-2-Nitrocinnamic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (E)-3-(2-Nitrophenyl)acrylic acid

A13171.14 A13171.22 A13171.36
A13171.14EA 67.8 EUR
A13171.14 A13171.22 A13171.36
trans-2-Nitrocinnamic acid ≥98%
trans-2-Nitrocinnamic acid

Purity: 98% CAS No.: 1013-96-3 Molecular Formula: C9H7NO4 Formula Weight: 193.16 Odour: Odourless Melting Point: 245-249° Beilstein Reference: 2211209 MDL No.: MFCD00007189

Formula: C₉H₇NO₄
MW: 193.16 g/mol
Melting Pt: 245…249 °C
Storage Temperature: Ambient
MDL Number: MFCD00007189
CAS Number: 1013-96-3
EINECS: 210-309-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR