2,3-Dimethyl-2-butene ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Tetramethylethylene

A13201.06
A13201.06EA 39.7 EUR
A13201.06
2,3-Dimethyl-2-butene ≥97%
2,3-Dimethyl-2-butene

Purity: 97% CAS No.: 563-79-1 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -17°(1°F) Refractive Index: 1.4120 Odour: Mild Melting Point: -75° Boiling Point: 72-73° Beilstein Reference: 1361357 UN No.: 3295 MDL No.: MFCD00008897

Formula: C₆H₁₂
Boiling Pt: 72…73 °C
Melting Pt: –75 °C
Density: 0,705
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008897
CAS Number: 563-79-1
EINECS: 209-263-8
UN: 3295
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR