3-Chlorobenzaldehyde ≥97%

Supplier: Thermo Fisher Scientific
Warning

A13254.14 A13254.22 A13254.36
A13254.14EA 44.6 EUR
A13254.14 A13254.22 A13254.36
3-Chlorobenzaldehyde ≥97%
3-Chlorobenzaldehyde

Purity: 97% CAS No.: 587-04-2 Molecular Formula: C7H5ClO Formula Weight: 140.57 Flash Point: 88°(190°F) Refractive Index: 1.5640 Odour: Characteristic Melting Point: 16-18° Boiling Point: 213-214° Beilstein Reference: 507098 MDL No.: MFCD00003350

Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 213…214 °C
Melting Pt: 16…18 °C
Density: 1.239
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR