Dimethyl sulphoxide ≥99%

Supplier: Thermo Fisher Scientific
Warning

A13280.36 A13280.0C A13280.0E
A13280.36EA 36.5 EUR
A13280.36 A13280.0C A13280.0E
Dimethyl sulphoxide ≥99%
Dimethyl sulphoxide

Purity: 99+% CAS No.: 67-68-5 Molecular Formula: C2H6OS Formula Weight: 78.13 Flash Point: 87°(189°F) Refractive Index: 1.4790 Odour: Mild Melting Point: 18.4° Boiling Point: 189° Beilstein Reference: 506008 Merck Reference: 14,3259 MDL No.: MFCD00002089

Formula: (CH₃)₂SO
Boiling Pt: 189 °C
Melting Pt: 18,4 °C
Density: 1,101
Flash Pt: 87 °C (189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 14,3259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR