3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥97%

Supplier: Thermo Fisher Scientific
Warning

A13449.06 A13449.14
A13449.06EA 54.6 EUR
A13449.06 A13449.14
3-Bromo-5-chloro-2-hydroxybenzaldehyde ≥97%
3-Bromo-5-chloro-2-hydroxybenzaldehyde

Purity: 97% CAS No.: 19652-32-5 Molecular Formula: C7H4BrClO2 Formula Weight: 235.47 Melting Point: 84-88° Beilstein Reference: 2362159

Formula: C₇H₄BrClO₂
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00051690
CAS Number: 19652-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR