2,4-Dihydroxybenzoic acid ≥97%

Supplier: Thermo Fisher Scientific
Warning

A13545.0E A13545.22 A13545.36
A13545.0EEA 331 EUR
A13545.0E A13545.22 A13545.36
2,4-Dihydroxybenzoic acid ≥97%
2,4-Dihydroxybenzoic acid

Purity: 97% CAS No.: 89-86-1 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 226° dec. Beilstein Reference: 1946213 Merck Reference: 14,8157 MDL No.: MFCD00002451

Formula: C₇H₆O₄
Boiling Pt: 390 °C (1013 hPa)
Melting Pt: ∼226 °C (dec.)
Density: 1,54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002451
CAS Number: 89-86-1
EINECS: 201-946-9
Merck Index: 14,8157

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR