Rhodamine B

Supplier: Thermo Fisher Scientific
Danger

A13572.0B A13572.18 A13572.30
A13572.0BEA 395 EUR
A13572.0B A13572.18 A13572.30
Rhodamine B
Rhodamine B

CAS No.: 81-88-9 Molecular Formula: C28H31ClN2O3 Formula Weight: 479.02 Physical Form: Crystals Density: 1.31 Appearance: Purple powder/dark green Melting Point: ca 210° dec. Beilstein Reference: 4119648 Merck Reference: 14,8183 MDL No.: MFCD00011931


Suitable for fluorescent labelling

Formula: C₂₈H₃₁ClN₂O₃
Melting Pt: ∼ 210 °C (dec.)
Density: 1,31
Storage Temperature: Ambient
MDL Number: MFCD00011931
CAS Number: 81-88-9
EINECS: 201-383-9
Merck Index: 14,8183

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR