2-Methoxy-p-cresol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A13603.09 A13603.18 A13603.30
A13603.09EA 33.3 EUR
A13603.09 A13603.18 A13603.30
2-Methoxy-p-cresol ≥98%
2-Methoxy-p-cresol

Purity: 98+% CAS No.: 93-51-6 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 99°(210°F) Refractive Index: 1.5370 Odour: Characteristic Melting Point: 5-6° Boiling Point: 220-222° Beilstein Reference: 1862340 Merck Reference: 14,2571 MDL No.: MFCD00002378

Formula: C₈H₁₀O₂
Boiling Pt: 220…222 °C
Melting Pt: 5…6 °C
Density: 1,092
Flash Pt: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002378
CAS Number: 93-51-6
EINECS: 202-252-9
Merck Index: 14,2571

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR