L(+)-Isoleucine ≥99%

Supplier: Thermo Fisher Scientific

A13699.09 A13699.18 A13699.22 A13699.30
A13699.09EA 36.3 EUR
A13699.09 A13699.18 A13699.22 A13699.30
L(+)-Isoleucine ≥99%
L(+)-Isoleucine

Purity: 99% CAS No.: 73-32-5 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: ca 287° dec. Beilstein Reference: 1721792 Merck Reference: 14,5179 Optical Rotation: +40° (c=5 in 5N HCl) MDL No.: MFCD00064222

Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Melting Pt: ∼ 287 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,5179

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR