2-Aminophenol 99%

Supplier: Thermo Fisher Scientific
Warning

A13735.22 A13735.36
A13735.22EA 35.2 EUR
A13735.22 A13735.36
2-Aminophenol 99%
2-Aminophenol

Purity: 99% CAS No.: 95-55-6 Molecular Formula: C6H7NO Formula Weight: 109.13 Flash Point: 175°(347°F) Odour: Characteristic Boiling Point: 150°/11mm Beilstein Reference: 606075 Merck Reference: 14,461 UN No.: 2512 MDL No.: MFCD00007690

Formula: C₆H₇NO
Boiling Pt: 150 °C (11 mmHg)
Melting Pt: 173…177 °C
Density: 1,328
Flash Pt: >175 °C (347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007690
CAS Number: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 14,461

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR