Palmitoyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A13812.22 A13812.36
A13812.22EA 78.2 EUR
A13812.22 A13812.36
Palmitoyl chloride ≥98%
Palmitoyl chloride

Purity: 98% CAS No.: 112-67-4 Molecular Formula: C16H31ClO Formula Weight: 274.88 Flash Point: 182°(359°F) Refractive Index: 1.4520 Melting Point: ca 9-12° Boiling Point: 174°/0.5mm Beilstein Reference: 972409 UN No.: 3265 MDL No.: MFCD00000742

Formula: C₁₆H₃₁ClO
Boiling Pt: 174 °C (0,5 mmHg)
Melting Pt: ∼ 9…12 °C
Density: 0,906
Flash Pt: 182 °C (359 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000742
CAS Number: 112-67-4
EINECS: 203-996-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR