(R)-(-)-Carvone ≥98%

Supplier: Thermo Fisher Scientific
Warning

A13900.0B A13900.18 A13900.30
A13900.0BEA 332 EUR
A13900.0B A13900.18 A13900.30
(R)-(-)-Carvone ≥98%
(R)-(-)-Carvone

Purity: 98% CAS No.: 6485-40-1 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 93°(199°F) Physical Form: Liquid Density: 0.959 Refractive Index: 1.4990 Appearance: Colourless Odour: Pleasant Boiling Point: 228-230° Beilstein Reference: 2206714 Optical Rotation: -61° (neat) MDL No.: MFCD00001578

Formula: C₁₀H₁₄O
Boiling Pt: 228…230 °C
Melting Pt: 25,2 °C
Density: 0,959
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001578
CAS Number: 6485-40-1
EINECS: 229-352-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR