Diphenyl diselenide ≥98%

Supplier: Thermo Fisher Scientific
Danger

A13919.06 A13919.14 A13919.22
A13919.06EA 53.5 EUR
A13919.06 A13919.14 A13919.22
Diphenyl diselenide ≥98%
Diphenyl diselenide

Purity: 98% CAS No.: 1666-13-3 Molecular Formula: C12H10Se2 Formula Weight: 312.13 Melting Point: 61-63° Beilstein Reference: 2047179 UN No.: 3283 MDL No.: MFCD00003001

Formula: C₁₂H₁₀Se₂
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 61…63 °C
Density: 1,32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003001
CAS Number: 1666-13-3
EINECS: 216-780-2
UN: 3283
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR