2-Methyl-1,3-cyclopentanedione ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 2-Methylcyclopentane-1,3-dione

A13936.06 A13936.14 A13936.22
A13936.06EA 54.2 EUR
A13936.06 A13936.14 A13936.22
2-Methyl-1,3-cyclopentanedione ≥98%
2-Methyl-1,3-cyclopentanedione

Purity: 98% CAS No.: 765-69-5 Molecular Formula: C6H8O2 Formula Weight: 112.13 Melting Point: 212-216° Beilstein Reference: 1237255

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 213
Melting Pt: 212…216 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001406
CAS Number: 765-69-5
EINECS: 212-153-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR