α,α'-Dibromo-p-xylene ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: p-Xylylene dibromide, 1,4-Bis(bromomethyl)benzene

A13939.09 A13939.18
A13939.09EA 39.4 EUR
A13939.09 A13939.18
α,α'-Dibromo-p-xylene ≥97%
α,α'-Dibromo-p-xylene

Purity: 97% CAS No.: 623-24-5 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 144-148° Boiling Point: 245° UN No.: 3261 MDL No.: MFCD00000182

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 245 °C
Melting Pt: 144…148 °C
Density: 2.012
Storage Temperature: Ambient
MDL Number: MFCD00000182
CAS Number: 623-24-5
EINECS: 210-781-1
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR