2-(Chloromethyl)quinoline hydrochloride ≥97%

Supplier: Thermo Fisher Scientific
Danger

A14102.03 A14102.06
A14102.03EA 33.7 EUR
A14102.03 A14102.06
2-(Chloromethyl)quinoline hydrochloride ≥97%
2-(Chloromethyl)quinoline hydrochloride

Purity: 97% CAS No.: 3747-74-8 Molecular Formula: C10H8ClN.HCl Formula Weight: 214.10 Melting Point: 184-190° Beilstein Reference: 3565832

Formula: C₁₀H₉Cl₂N
MW: 214.10 g/mol
Melting Pt: 184…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012734
CAS Number: 3747-74-8
EINECS: 223-145-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR