Benzyltriethylammonium bromide ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14291.14 A14291.22 A14291.36
A14291.14EA 33.3 EUR
A14291.14 A14291.22 A14291.36
Benzyltriethylammonium bromide ≥98%
Benzyltriethylammonium bromide

Purity: 98+% CAS No.: 5197-95-5 Molecular Formula: (CH3CH2)3N(CH2C6H5)Br Formula Weight: 272.24 Melting Point: ca 200° dec. Beilstein Reference: 3739113

Formula: C₁₃H₂₂BrN
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011822
CAS Number: 5197-95-5
EINECS: 225-986-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR