6-Azauracil ≥98%

Supplier: Thermo Fisher Scientific
Danger

A14389.14 A14389.06
A14389.14EA 206 EUR
A14389.14 A14389.06
6-Azauracil ≥98%
6-Azauracil

Purity: 98% CAS No.: 461-89-2 Molecular Formula: C3H3N3O2 Formula Weight: 113.08 Melting Point: 274-282° Beilstein Reference: 116472

Formula: C₃H₃N₃O₂
Melting Pt: 274…282 °C
Storage Temperature: Ambient
MDL Number: MFCD00006456
CAS Number: 461-89-2
EINECS: 207-318-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR