2,2-Dimethyl-1-propanol ≥99%

Supplier: Thermo Fisher Scientific
Danger

A14390.14 A14390.22 A14390.36
A14390.14EA 40.1 EUR
A14390.14 A14390.22 A14390.36
2,2-Dimethyl-1-propanol ≥99%
2,2-Dimethyl-1-propanol

Purity: 99% CAS No.: 75-84-3 Molecular Formula: (CH3)3CCH2OH Formula Weight: 88.15 Flash Point: 36°(96°F) Melting Point: 52-56° Boiling Point: 111-114° Beilstein Reference: 1730984 Merck Reference: 14,6457 UN No.: 1325 MDL No.: MFCD00004682

Formula: C₅H₁₂O
Boiling Pt: 111…114 °C
Melting Pt: 52…56 °C
Density: 0,818
Flash Pt: 36 °C (96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004682
CAS Number: 75-84-3
EINECS: 200-907-3
UN: 1325
ADR: 4.1,II
Merck Index: 14,6457

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR