Ethyl nitroacetate ≥97%

Supplier: Thermo Fisher Scientific
Warning

A14433.06 A14433.14 A14433.22
A14433.06EA 65.1 EUR
A14433.06 A14433.14 A14433.22
Ethyl nitroacetate ≥97%
Ethyl nitroacetate

Purity: 97% CAS No.: 626-35-7 Molecular Formula: C4H7NO4 Formula Weight: 133.11 Flash Point: 92°(197°F) Refractive Index: 1.4240 Odour: Weak Boiling Point: 93-95°/10mm Beilstein Reference: 1210027 MDL No.: MFCD00007403

Formula: C₄H₇NO₄
Boiling Pt: 93…95 °C (10 mmHg)
Density: 1,202
Flash Pt: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007403
CAS Number: 626-35-7
EINECS: 210-944-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR