2-Chloroethylammonium chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

A14455.30 A14455.0B A14455.18
A14455.30EA 118 EUR
A14455.30 A14455.0B A14455.18
2-Chloroethylammonium chloride ≥98%
2-Chloroethylammonium chloride

Purity: 98+% CAS No.: 870-24-6 Molecular Formula: ClCH2CH2NH2.HCl Formula Weight: 115.99 Odour: Odourless pH: 2-3 Melting Point: 143-148° Beilstein Reference: 3590304 UN No.: 3261 MDL No.: MFCD00012887

Formula: C₂H₇Cl₂N
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00012887
CAS Number: 870-24-6
EINECS: 212-793-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR