α,α,α-Trifluoro-m-toluoyl chloride ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride

A14493.06 A14493.14 A14493.22
A14493.06EA 50.2 EUR
A14493.06 A14493.14 A14493.22
α,α,α-Trifluoro-m-toluoyl chloride ≥97%
α,α,α-Trifluoro-m-toluoyl chloride

Purity: 97% CAS No.: 2251-65-2 Molecular Formula: C8H4ClF3O Formula Weight: 208.57 Flash Point: 110°(230°F) Refractive Index: 1.4770 Odour: Pungent Boiling Point: 189-191° Beilstein Reference: 391266 UN No.: 3265 MDL No.: MFCD00000680

Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
Boiling Pt: 189…191 °C
Density: 1.408
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000680
CAS Number: 2251-65-2
EINECS: 218-844-5
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR