Flavin adenine dinucleotide disodium salt hydrate, (max. 10% H₂O) 94% (dry weight)

Supplier: Thermo Fisher Scientific

Synonyms: FAD-Na2 hydrate, FAD disodium salt hydrate, Riboflavin 5'-adenosine diphosphate disodium salt hydrate, Flavin Adenine Dinucleotide DiSodium Salt xhydrate

A14495.MEEA 194 EUR
A14495.ME A14495.MC
Flavin adenine dinucleotide disodium salt hydrate, (max. 10% H₂O) 94% (dry weight)
Flavin adenine dinucleotide disodium salt hydrate

Purity: 94% CAS No.: 84366-81-4 Molecular Formula: C27H31N9Na2O15P2.xH2O Formula Weight: 829.52 Storage Temperature: 2°C to 8°C MDL No.: MFCD00151217

Formula: C₂₇H₃₃N₉Na₂O₁₆P₂
Storage Temperature: Refrigerator
MDL Number: MFCD00151217
CAS Number: 1891059-93-0
EINECS: 282-733-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR