(4-Pyridyl)acetic acid ethyl ester ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14550.22 A14550.03 A14550.06 A14550.14
A14550.22EA 1260 EUR
A14550.22 A14550.03 A14550.06 A14550.14
(4-Pyridyl)acetic acid ethyl ester ≥98%
(4-Pyridyl)acetic acid ethyl ester

Purity: 98% CAS No.: 54401-85-3 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 110°(230°F) Refractive Index: 1.500 Melting Point: 18-20° Boiling Point: 86-88°/1mm Beilstein Reference: 123786 MDL No.: MFCD00051830

Formula: C₉H₁₁NO₂
Boiling Pt: 86…88 °C (1 mmHg)
Melting Pt: 18…20 °C
Density: 1,080
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051830
CAS Number: 54401-85-3
EINECS: 259-150-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR