4-tert-Butylpyrocatechol ≥98%

Supplier: Thermo Fisher Scientific
Danger

A14599.0B A14599.0I A14599.30
A14599.0BEA 147 EUR
A14599.0B A14599.0I A14599.30
4-tert-Butylpyrocatechol ≥98%
4-tert-Butylpyrocatechol

Purity: 98% CAS No.: 98-29-3 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 151°(303°F) Odour: Odourless Boiling Point: 284-286° Beilstein Reference: 2043335 UN No.: 2923 MDL No.: MFCD00002201

Formula: C₁₀H₁₄O₂
Boiling Pt: 284…286 °C
Melting Pt: 53…57 °C
Density: 1,049
Flash Pt: 151 °C (303 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002201
CAS Number: 98-29-3
EINECS: 202-653-9
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR