Sodium trifluoroacetate ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14613.14 A14613.22 A14613.36
A14613.14EA 44 EUR
A14613.14 A14613.22 A14613.36
Sodium trifluoroacetate ≥98%
Sodium trifluoroacetate

Purity: 98% CAS No.: 2923-18-4 Molecular Formula: CF3CO2Na Formula Weight: 136.00 Melting Point: ca 207° dec. Beilstein Reference: 3597949

Formula: C₂F₃NaO₂
Melting Pt: ∼ 207 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013217
CAS Number: 2923-18-4
EINECS: 220-879-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR