1,4-Dithiane-2,5-diol 96%

Supplier: Thermo Fisher Scientific
Danger

A14620.0B A14620.18 A14620.30
A14620.0BEA 324 EUR
A14620.0B A14620.18 A14620.30
1,4-Dithiane-2,5-diol 96%
1,4-Dithiane-2,5-diol

Purity: 96% CAS No.: 40018-26-6 Molecular Formula: C4H8O2S2 Formula Weight: 152.24 Storage Temperature: 2°C to 8°C Melting Point: ca 130° dec. Beilstein Reference: 104402 UN No.: 2811 MDL No.: MFCD00006659

Formula: C₄H₈O₂S₂
Melting Pt: ∼ 130 °C (dec.)
MDL Number: MFCD00006659
CAS Number: 40018-26-6
EINECS: 254-751-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR