Cyclopentyltriphenylphosphonium bromide 98

Supplier: Thermo Fisher Scientific
Warning

A14647.09 A14647.18 A14647.30
A14647.09EA 46.2 EUR
A14647.09 A14647.18 A14647.30
Cyclopentyltriphenylphosphonium bromide 98
Cyclopentyltriphenylphosphonium bromide

Purity: 98% CAS No.: 7333-52-0 Molecular Formula: C23H24BrP Formula Weight: 411.33 Melting Point: 262-266° Beilstein Reference: 3580193

Formula: C₂₃H₂₄BrP
MW: 411.32 g/mol
Melting Pt: 262…266 °C
Storage Temperature: Ambient
MDL Number: MFCD00051880
CAS Number: 7333-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR