3-Chloro-4-iodobenzotrifluoride ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14685.03 A14685.06
A14685.03EA 0 EUR
A14685.03 A14685.06
3-Chloro-4-iodobenzotrifluoride ≥98%
3-Chloro-4-iodobenzotrifluoride

Purity: 98% CAS No.: 141738-80-9 Molecular Formula: C7H3ClF3I Formula Weight: 306.45 Boiling Point: 58-60°/0.2mm MDL No.: MFCD00042562

Formula: C₇H₃ClF₃I
Boiling Pt: 58…60 °C (0,2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00042562
CAS Number: 141738-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR