Tricine ≥98%

Supplier: Thermo Fisher Scientific

A14695.14 A14695.22 A14695.36
A14695.14EA 30.5 EUR
A14695.14 A14695.22 A14695.36
Tricine ≥98%
Tricine

Purity: 98+% CAS No.: 5704-04-1 Molecular Formula: (HOCH2)3CNHCH2CO2H Formula Weight: 179.17 Melting Point: ca 183° dec. Beilstein Reference: 1937804 Merck Reference: 14,9651 MDL No.: MFCD00004277

Formula: (HOCH₂)₃CNHCH₂CO₂H
Melting Pt: ∼ 183 °C (dec.)
Density: 1,523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 14,9651

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR