n-Decane 99

Supplier: Thermo Fisher Scientific
Danger

A14732.0F A14732.AE A14732.AP
A14732.0FEA 586 EUR
A14732.0F A14732.AE A14732.AP
n-Decane 99
n-Decane

Purity: 99% CAS No.: 124-18-5 Molecular Formula: C10H22 Formula Weight: 142.29 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.730 Refractive Index: 1.4115 Appearance: Colourless Odour: Petrol-like Melting Point: -30° Boiling Point: 172-174° Solubility: Soluble in alcohol and ethers Beilstein Reference: 1696981 UN No.: 2247 MDL No.: MFCD00008954

Formula: H₃C(CH₂)₈CH₃
MW: 142.28 g/mol
Boiling Pt: 172…174 °C
Melting Pt: –30 °C
Density: 0.730
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
EINECS: 204-686-4
UN: 2247
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR