Calcium citrate tetrahydrate 96%

Supplier: Thermo Fisher Scientific

A14828.22 A14828.36
A14828.22EA 28.6 EUR
A14828.22 A14828.36
Calcium citrate tetrahydrate 96%
Calcium citrate tetrahydrate

Purity: 96% CAS No.: 5785-44-4 Molecular Formula: C12H10Ca3O14.4H2O Formula Weight: 572.60 (500.54anhy) MDL No.: MFCD00150786

Formula: C₁₂H₁₀Ca₃O₁₄·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150786
CAS Number: 5785-44-4
EINECS: 212-391-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR