Hydroquinone monomethyl ether ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole

A14845.0B A14845.18 A14845.30
A14845.0BEA 130 EUR
A14845.0B A14845.18 A14845.30
Hydroquinone monomethyl ether ≥98%
Hydroquinone monomethyl ether

Purity: 98+% CAS No.: 150-76-5 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 132°(269°F) Melting Point: 55-58° Boiling Point: 243-246° Beilstein Reference: 507924 MDL No.: MFCD00002332

Formula: C₇H₈O₂
Boiling Pt: 243…246 °C
Melting Pt: 55…58 °C
Density: 1,550
Flash Pt: 132 °C (269 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5
EINECS: 205-769-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR