3-Bromophenol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14849.09 A14849.18 A14849.30
A14849.09EA 36.6 EUR
A14849.09 A14849.18 A14849.30
3-Bromophenol ≥98%
3-Bromophenol

Purity: 98% CAS No.: 591-20-8 Molecular Formula: C6H5BrO Formula Weight: 173.01 Flash Point: 110°(230°F) Refractive Index: 1.5972 Melting Point: 28-32° Boiling Point: 235-236° Beilstein Reference: 1853950 Merck Reference: 14,1428 MDL No.: MFCD00002253

Formula: C₆H₅BrO
Boiling Pt: 235…236 °C
Melting Pt: 28…32 °C
Density: 1,63
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002253
CAS Number: 591-20-8
EINECS: 209-706-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,1428

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR