2-Bromo-5-fluorophenol ≥97%

Supplier: Thermo Fisher Scientific
Danger

A15064.06 A15064.14
A15064.06EA 42.9 EUR
A15064.06 A15064.14
2-Bromo-5-fluorophenol ≥97%
2-Bromo-5-fluorophenol

Purity: 97% CAS No.: 147460-41-1 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Flash Point: 87°(188°F) Refractive Index: 1.5540 Boiling Point: 184-186° Beilstein Reference: 7915214 MDL No.: MFCD00040939

Formula: C₆H₄BrFO
Boiling Pt: 184…186 °C
Density: 1,717
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040939
CAS Number: 147460-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR