5-Iodovanillin ≥98%

Supplier: Thermo Fisher Scientific
Warning

A15162.09 A15162.18 A15162.30
A15162.09EA 33.6 EUR
A15162.09 A15162.18 A15162.30
5-Iodovanillin ≥98%
5-Iodovanillin

Purity: 98% CAS No.: 5438-36-8 Molecular Formula: C8H7IO3 Formula Weight: 278.05 Odour: Irritating Melting Point: 181-184° Beilstein Reference: 2049762 MDL No.: MFCD00006941

Formula: C₈H₇IO₃
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006941
CAS Number: 5438-36-8
EINECS: 226-617-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR