9-Acridone ≥99%

Supplier: Thermo Fisher Scientific

A15222.06 A15222.14
A15222.06EA 67 EUR
A15222.06 A15222.14
9-Acridone ≥99%
9-Acridone

Purity: 99% CAS No.: 578-95-0 Molecular Formula: C13H9NO Formula Weight: 195.22 Melting Point: 360-362° Beilstein Reference: 7104

Formula: C₁₃H₉NO
Melting Pt: 360…362 °C
Storage Temperature: Ambient
MDL Number: MFCD00005019
CAS Number: 578-95-0
EINECS: 209-434-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR