3-Cyclohexene-1-carboxylic acid 98%

Supplier: Thermo Fisher Scientific
Danger

A15229.06 A15229.14
A15229.06EA 80.4 EUR
A15229.06 A15229.14
3-Cyclohexene-1-carboxylic acid 98%
3-Cyclohexene-1-carboxylic acid

Purity: 98% CAS No.: 4771-80-6 Molecular Formula: C7H10O2 Formula Weight: 126.16 Flash Point: 110°(230°F) Refractive Index: 1.4815 Melting Point: 16-18° Boiling Point: 120-122°/10mm Solubility: V sol Beilstein Reference: 1617723 UN No.: UN3265 MDL No.: MFCD00013781

Formula: C₇H₁₀O₂
Boiling Pt: 120…122 °C (10 mmHg)
Melting Pt: 16…18 °C
Density: 1,082
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013781
CAS Number: 4771-80-6
EINECS: 225-314-7
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR