Methyl-3-hydroxy-4-nitrobenzoate ≥98%

Supplier: Thermo Fisher Scientific
Warning

A15329.06 A15329.14 A15329.22
A15329.06EA 49.9 EUR
A15329.06 A15329.14 A15329.22
Methyl-3-hydroxy-4-nitrobenzoate ≥98%
Methyl 3-hydroxy-4-nitrobenzoate

Purity: 98% CAS No.: 713-52-0 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 89-92° Beilstein Reference: 2583832

Formula: C₈H₇NO₅
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00016997
CAS Number: 713-52-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR