Creatine phosphate disodium salt tetrahydrate ≥98%

Supplier: Thermo Fisher Scientific

A15362.03 A15362.06 A15362.14
A15362.03EA 32.6 EUR
A15362.03 A15362.06 A15362.14
Creatine phosphate disodium salt tetrahydrate ≥98%
Creatine phosphate disodium salt tetrahydrate

Purity: 98+% CAS No.: 71519-72-7 Molecular Formula: C4H8N3Na2O5P.4H2O Formula Weight: 327.14 (255.07anhy) Storage Temperature: 2°C to 8°C


Substrate for creatine kinase determinations

Formula: Na₂O₃PNHC(=NH)N(CH₃)CH₂COOH·4H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150192
CAS Number: 71519-72-7
EINECS: 213-074-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR