3,5-Bis(trifluoromethyl)benzonitrile ≥98%

Supplier: Thermo Fisher Scientific
Warning

A15460.06 A15460.14
A15460.06EA 73 EUR
A15460.06 A15460.14
3,5-Bis(trifluoromethyl)benzonitrile ≥98%
3,5-Bis(trifluoromethyl)benzonitrile

Purity: 98% CAS No.: 27126-93-8 Molecular Formula: C9H3F6N Formula Weight: 239.12 Flash Point: 72°(161°F) Refractive Index: 1.4175 Melting Point: 16-20° Boiling Point: 154-156° Beilstein Reference: 3552650 UN No.: 3276 MDL No.: MFCD00000379

Formula: C₉H₃F₆N
Boiling Pt: 154…156 °C
Melting Pt: 16…20 °C
Density: 1,421
Flash Pt: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000379
CAS Number: 27126-93-8
EINECS: 248-240-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR