D-(+)-Glucosamine hydrochloride ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

A15532.0B A15532.18 A15532.30
A15532.0BEA 389 EUR
A15532.0B A15532.18 A15532.30
D-(+)-Glucosamine hydrochloride ≥98%
D-(+)-Glucosamine hydrochloride

Purity: 98+% CAS No.: 66-84-2 Molecular Formula: C6H13NO5.HCl Formula Weight: 215.64 Melting Point: ca 192° dec. Beilstein Reference: 4157370 Optical Rotation: +71.5° (c=1 in water, 20h) MDL No.: MFCD00067674

Formula: C₆H₁₃NO₅
MW: 215.63 g/mol
Melting Pt: ∼ 192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00135831
CAS Number: 66-84-2
EINECS: 200-638-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR