Polyethylene oxide, M.W. >5,000,000

Supplier: Thermo Fisher Scientific

Synonyms: PEO

A15536.22 A15536.36
A15536.22EA 140 EUR
A15536.22 A15536.36
Polyethylene oxide, M.W. >5,000,000
Polyethylene oxide

CAS No.: 25322-68-3 Molecular Formula: (-CH2CH2O-)n Odour: Odourless pH: 5-7 Melting Point: ca 175° MDL No.: MFCD00147992

Formula: (-CH₂CH₂O-)n
Melting Pt: ∼ 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00147970
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR