Propylene carbonate 99% (by GC)

Supplier: Thermo Fisher Scientific
Warning

A15552.30 A15552.0B A15552.A4 A15552.XL
A15552.30EA 26.2 EUR
A15552.30 A15552.0B A15552.A4 A15552.XL
Propylene carbonate 99% (by GC)
Propylene carbonate

Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries.


It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.


Caution: Moisture sensitive. Hygroscopic. Incompatible with acids and bases.

Formula: C₄H₆O₃
Boiling Pt: 240…243 °C
Melting Pt: –49 °C
Density: 1,204
Flash Pt: 132 °C (270 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR