Uracil ≥99%

Supplier: Thermo Fisher Scientific

A15570.0B A15570.18 A15570.30
A15570.0BEA 664 EUR
A15570.0B A15570.18 A15570.30
Uracil ≥99%
Uracil

Purity: 99+% CAS No.: 66-22-8 Molecular Formula: C4H4N2O2 Formula Weight: 112.09 Physical Form: Powder Appearance: White Melting Point: ca 330° Beilstein Reference: 606623 Merck Reference: 14,9850 MDL No.: MFCD00006016

Formula: C₄H₄N₂O₂
Boiling Pt: 379 °C (1013 hPa)
Melting Pt: ∼ 330 °C
Density: 0,99744 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9
Merck Index: 14,9850

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR